Assay Details
| Gene Symbol: AUNIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544385.2 | XP_005544442.1 | 2 - 3 | 313 | 151 | |
| XM_005544386.2 | XP_005544443.1 | 2 - 3 | 835 | 151 | ||
| XM_005544387.2 | XP_005544444.1 | 2 - 3 | 574 | 151 | ||
| XM_005544388.2 | XP_005544445.1 | 3 - 4 | 476 | 151 | ||
| XM_015440533.1 | XP_015296019.1 | 3 - 4 | 583 | 151 | ||
| XM_015440546.1 | XP_015296032.1 | 2 - 3 | 492 | 151 | ||
More Information
Set Membership: |
|

AUNIP
Genomic Map