Assay Details
| Gene Symbol: TNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554316.1 | XP_005554373.1 | 5 - 6 | 1086 | 166 | |
| XM_005554317.1 | XP_005554374.1 | 4 - 5 | 837 | 166 | ||
More Information
Set Membership: |
|

TNIP2
Genomic Map