Assay Details
| Gene Symbol: RIPOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592271.2 | XP_005592328.1 | 3 - 4 | 582 | 122 | |
| XM_015443013.1 | XP_015298499.1 | 2 - 3 | 628 | 122 | ||
| XM_015443014.1 | XP_015298500.1 | 2 - 3 | 628 | 122 | ||
| XM_015443015.1 | XP_015298501.1 | 2 - 3 | 246 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015443015.1 transcripts.
Set Membership: |
|

RIPOR1
Genomic Map