Assay Details
Gene Symbol: KDM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591505.2 | XP_005591562.1 | 2 - 3 | 1540 | 137 | |
XM_005591506.2 | XP_005591563.1 | 2 - 3 | 2162 | 137 | ||
XM_005591507.2 | XP_005591564.1 | 2 - 3 | 2059 | 137 | ||
XM_005591509.2 | XP_005591566.1 | 2 - 3 | 1540 | 137 | ||
XM_005591510.2 | XP_005591567.1 | 2 - 3 | 1540 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
