Assay Details
Gene Symbol: RABEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591543.2 | XP_005591600.1 | 4 - 5 | 1125 | 80 | |
XM_005591544.2 | XP_005591601.1 | 4 - 5 | 506 | 80 | ||
XM_015442503.1 | XP_015297989.1 | 5 - 6 | 707 | 80 | ||
GenBank mRNA | AB169254.1 | - | 4 - 5 | 506 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
