Assay Details
Gene Symbol: RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553651.2 | XP_005553708.1 | 3 - 4 | 316 | 123 | |
XM_005553652.2 | XP_005553709.1 | 3 - 4 | 317 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
