Assay Details
| Gene Symbol: MAP2K2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587544.2 | XP_005587601.1 | 1 - 2 | 404 | 117 | |
| XM_005587545.2 | XP_005587602.1 | 1 - 2 | 404 | 117 | ||
More Information
Set Membership: |
|

MAP2K2
Genomic Map