Assay Details
Gene Symbol: CPLANE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544674.2 | XP_005544731.1 | 5 - 6 | 808 | 125 | |
XM_015442204.1 | XP_015297690.1 | 4 - 5 | 565 | 125 | ||
XM_015442208.1 | XP_015297694.1 | 5 - 6 | 1163 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
