Assay Details
| Gene Symbol: SLCO5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563500.2 | XP_005563557.1 | 9 - 10 | 2780 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLCO5A1| Gene Symbol: SLCO5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563500.2 | XP_005563557.1 | 9 - 10 | 2780 | 110 | |
Set Membership: |
|