Assay Details
| Gene Symbol: GALNTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287656.1 | NP_001274585.1 | 5 - 6 | 875 | 58 | |
| XM_005551225.2 | XP_005551282.1 | 6 - 7 | 1018 | 58 | ||
| GenBank mRNA | AB069985.1 | - | 6 - 7 | 1038 | 58 | |
| AB070053.1 | - | 5 - 6 | 875 | 58 | ||
More Information
Set Membership: |
|

GALNTL5
Genomic Map