Assay Details
Gene Symbol: ZSCAN21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549164.2 | XP_005549221.1 | 3 - 4 | 770 | 93 | |
XM_015447130.1 | XP_015302616.1 | 3 - 4 | 708 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
