Assay Details
| Gene Symbol: LACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585759.2 | XP_005585816.1 | 3 - 4 | 878 | 90 | |
| XM_005585760.2 | XP_005585817.1 | 3 - 4 | 882 | 90 | ||
More Information
Set Membership: |
|

LACC1
Genomic Map