Assay Details
| Gene Symbol: KSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572357.2 | XP_005572414.1 | 4 - 5 | 1188 | 85 | |
| XM_005572358.2 | XP_005572415.1 | 4 - 5 | 1188 | 85 | ||
| XM_015431615.1 | XP_015287101.1 | 4 - 5 | 1188 | 85 | ||
| XM_015431616.1 | XP_015287102.1 | 4 - 5 | 2327 | 85 | ||
More Information
Set Membership: |
|

KSR2
Genomic Map