Assay Details
Gene Symbol: LOC102121963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005543926.2 | XP_005543983.1 | 2 - 3 | 256 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
