Assay Details
Gene Symbol: LOC102137734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592150.2 | XP_005592207.1 | 1 - 2 | 229 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
