Assay Details
Gene Symbol: CFAP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005547908.2 | XP_005547965.1 | 5 - 6 | 950 | 103 | |
GenBank mRNA | AB168339.1 | - | 5 - 6 | 527 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
