Assay Details
| Gene Symbol: SLC16A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565795.2 | XP_005565852.1 | 4 - 5 | 887 | 93 | |
| XM_005565796.2 | XP_005565853.1 | 4 - 5 | 1438 | 93 | ||
| XM_015456344.1 | XP_015311830.1 | 4 - 5 | 1570 | 93 | ||
| XM_015456345.1 | XP_015311831.1 | 4 - 5 | 1331 | 93 | ||
| XM_015456346.1 | XP_015311832.1 | 3 - 4 | 2950 | 93 | ||
More Information
Set Membership: |
|

SLC16A9
Genomic Map