Assay Details
| Gene Symbol: BRINP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540259.2 | XP_005540316.1 | 7 - 8 | 1945 | 68 | |
| XM_005540260.2 | XP_005540317.1 | 7 - 8 | 2857 | 68 | ||
| XM_005540261.2 | XP_005540318.1 | 7 - 8 | 2584 | 68 | ||
| XM_015447231.1 | XP_015302717.1 | 6 - 7 | 1758 | 68 | ||
| XM_015447232.1 | XP_015302718.1 | 6 - 7 | 1283 | 68 | ||
| XM_015447239.1 | XP_015302725.1 | 6 - 7 | 2393 | 68 | ||
More Information
Set Membership: |
|

BRINP3
Genomic Map