Assay Details
Gene Symbol: ZNF790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015441084.1 | XP_015296570.1 | 4 - 5 | 592 | 78 | |
XM_015441085.1 | XP_015296571.1 | 4 - 5 | 1242 | 78 | ||
XM_015441086.1 | XP_015296572.1 | 4 - 5 | 389 | 78 | ||
XM_015441087.1 | XP_015296573.1 | 4 - 5 | 385 | 78 | ||
XM_015441088.1 | XP_015296574.1 | 4 - 5 | 1044 | 78 | ||
XM_015441089.1 | XP_015296575.1 | 4 - 5 | 1097 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
