Assay Details
Gene Symbol: TMEM207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005545411.2 | XP_005545468.1 | 4 - 5 | 499 | 83 | |
XM_005545412.1 | XP_005545469.1 | 4 - 5 | 390 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
