Assay Details
| Gene Symbol: GNAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587197.2 | XP_005587254.1 | 8 - 9 | 6905 | 97 | |
| XM_005587198.2 | XP_005587255.1 | 6 - 7 | 1350 | 97 | ||
| XM_005587199.2 | XP_005587256.1 | 6 - 7 | 430 | 97 | ||
More Information
Set Membership: |
|

GNAL
Genomic Map