Assay Details
| Gene Symbol: CALHM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566344.2 | XP_005566401.1 | 1 - 2 | 683 | 134 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CALHM1| Gene Symbol: CALHM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566344.2 | XP_005566401.1 | 1 - 2 | 683 | 134 | |
Set Membership: |
|