Assay Details
| Gene Symbol: CRAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590871.2 | XP_005590928.1 | 9 - 10 | 1811 | 52 | |
| XM_005590872.2 | XP_005590929.1 | 9 - 10 | 1809 | 52 | ||
| XM_005590873.2 | XP_005590930.1 | 9 - 10 | 1746 | 52 | ||
| XM_015442478.1 | XP_015297964.1 | 9 - 10 | 1814 | 52 | ||
| XM_015442479.1 | XP_015297965.1 | 9 - 10 | 1812 | 52 | ||
| XM_015442480.1 | XP_015297966.1 | 9 - 10 | 1383 | 52 | ||
| XM_015442481.1 | XP_015297967.1 | 9 - 10 | 1812 | 52 | ||
| XM_015442483.1 | XP_015297969.1 | 9 - 10 | 1816 | 52 | ||
| XM_015442484.1 | XP_015297970.1 | 9 - 10 | 1816 | 52 | ||
More Information
Set Membership: |
|

CRAMP1
Genomic Map