Assay Details
Gene Symbol: HMGB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283646.1 | NP_001270575.1 | 4 | 1787 | 85 | |
GenBank mRNA | AB168641.1 | - | 4 | 1787 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001283646.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
