Assay Details
| Gene Symbol: USF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548122.2 | XP_005548179.1 | 6 - 7 | 663 | 129 | |
| XM_015446405.1 | XP_015301891.1 | 5 - 6 | 985 | 129 | ||
| XM_015446406.1 | XP_015301892.1 | 5 - 6 | 545 | 129 | ||
More Information
Set Membership: |
|

USF3
Genomic Map