Assay Details
Gene Symbol: PGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590957.2 | XP_005591014.1 | 2 - 3 | 679 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
