Assay Details
| Gene Symbol: INSM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015457687.1 | XP_015313173.1 | 1 | 316 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015457687.1 transcripts.
Set Membership: |
|

INSM1
Genomic Map