Assay Details
Gene Symbol: KCNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569870.2 | XP_005569927.1 | 1 | 2589 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
