Assay Details
Gene Symbol: CAVIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005573792.2 | XP_005573849.1 | 1 - 2 | 851 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
