Assay Details
| Gene Symbol: KCNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542403.2 | XP_005542460.1 | 3 | 2956 | 131 | |
| XM_005542404.2 | XP_005542461.1 | 3 | 2395 | 131 | ||
| XM_005542405.2 | XP_005542462.1 | 3 | 2132 | 131 | ||
More Information
Set Membership: |
|

KCNA2
Genomic Map