Assay Details
| Gene Symbol: NFIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015436574.1 | XP_015292060.1 | 1 | 1263 | 152 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NFIL3| Gene Symbol: NFIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015436574.1 | XP_015292060.1 | 1 | 1263 | 152 | |
Set Membership: |
|