Assay Details
Gene Symbol: HOXA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549921.2 | XP_005549978.1 | 1 - 2 | 788 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
