Assay Details
| Gene Symbol: TOPORS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581555.2 | XP_005581612.1 | 2 - 3 | 560 | 159 | |
| GenBank mRNA | AB171977.1 | - | 2 - 3 | 329 | 159 | |
More Information
Set Membership: |
|

TOPORS
Genomic Map