Assay Details
Gene Symbol: LSM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005556021.2 | XP_005556078.1 | 2 - 3 | 232 | 131 | |
XM_005556022.2 | XP_005556079.1 | 3 - 4 | 325 | 131 | ||
XM_005556024.2 | XP_005556081.1 | 2 - 3 | 194 | 131 | ||
XM_015450883.1 | XP_015306369.1 | 3 - 4 | 331 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
