Assay Details
Gene Symbol: ZNF146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588944.2 | XP_005589001.1 | 3 | 286 | 184 | |
XM_005588946.2 | XP_005589003.1 | 2 | 223 | 184 | ||
XM_005588947.2 | XP_005589004.1 | 3 | 293 | 184 | ||
XM_005588948.2 | XP_005589005.1 | 2 | 215 | 184 | ||
XM_005588949.2 | XP_005589006.1 | 2 | 138 | 184 | ||
XM_015441666.1 | XP_015297152.1 | 3 | 297 | 184 | ||
XM_015441667.1 | XP_015297153.1 | 4 | 371 | 184 | ||
XM_015441669.1 | XP_015297155.1 | 4 | 368 | 184 | ||
XM_015441670.1 | XP_015297156.1 | 4 | 367 | 184 | ||
XM_015441671.1 | XP_015297157.1 | 4 | 364 | 184 | ||
XM_015441672.1 | XP_015297158.1 | 3 | 208 | 184 | ||
XM_015441673.1 | XP_015297159.1 | 3 | 205 | 184 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005588944.2, XM_005588946.2, XM_005588947.2, XM_005588948.2, XM_005588949.2, XM_015441666.1, XM_015441667.1, XM_015441669.1, XM_015441670.1, XM_015441671.1, XM_015441672.1, XM_015441673.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
