Assay Details
Gene Symbol: RAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005586557.2 | XP_005586614.1 | 2 - 3 | 766 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
