Assay Details
Gene Symbol: UBQLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015443625.1 | XP_015299111.1 | 1 | 798 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015443625.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
