Assay Details
Gene Symbol: MRPS28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005563600.2 | XP_005563657.2 | 2 - 3 | 419 | 123 | |
XM_005563601.2 | XP_005563658.2 | 2 - 3 | 406 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
