Assay Details
| Gene Symbol: YY1AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541526.2 | XP_005541583.1 | 4 - 5 | 568 | 134 | |
| XM_005541528.2 | XP_005541585.1 | 4 - 5 | 568 | 134 | ||
| XM_005541529.2 | XP_005541586.1 | 4 - 5 | 568 | 134 | ||
| GenBank mRNA | AB171182.1 | - | 4 - 5 | 568 | 134 | |
More Information
Set Membership: |
|

YY1AP1
Genomic Map