Assay Details
Gene Symbol: TCAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005551061.2 | XP_005551118.1 | 7 - 8 | 3072 | 86 | |
XM_005551062.2 | XP_005551119.1 | 7 - 8 | 2836 | 86 | ||
XM_005551063.2 | XP_005551120.1 | 7 - 8 | 2544 | 86 | ||
XM_005551064.2 | XP_005551121.1 | 6 - 7 | 2899 | 86 | ||
XM_005551065.2 | XP_005551122.1 | 6 - 7 | 2442 | 86 | ||
XM_005551066.2 | XP_005551123.1 | 6 - 7 | 2662 | 86 | ||
XM_005551067.2 | XP_005551124.1 | 6 - 7 | 2333 | 86 | ||
XM_015448200.1 | XP_015303686.1 | 7 - 8 | 3076 | 86 | ||
XM_015448201.1 | XP_015303687.1 | 6 - 7 | 2333 | 86 | ||
GenBank mRNA | AB173120.1 | - | 2 - 3 | 352 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
