Assay Details
Gene Symbol: LOC102119711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570923.1 | XP_005570980.1 | 8 - 9 | 1353 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102119711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570923.1 | XP_005570980.1 | 8 - 9 | 1353 | 140 |
Set Membership: |
![]() ![]() ![]() ![]() |