Assay Details
| Gene Symbol: PAFAH1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579740.2 | XP_005579797.1 | 2 - 3 | 229 | 91 | |
| XM_005579741.1 | XP_005579798.1 | 2 - 3 | 461 | 91 | ||
| XM_015435755.1 | XP_015291241.1 | 2 - 3 | 228 | 91 | ||
| XM_015435756.1 | XP_015291242.1 | 1 - 2 | 83 | 91 | ||
| GenBank mRNA | AB173767.1 | - | 2 - 3 | 466 | 91 | |
More Information
Set Membership: |
|

PAFAH1B2
Genomic Map