Assay Details
Gene Symbol: PPY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005584386.2 | XP_005584443.1 | 3 - 4 | 310 | 129 | |
XM_005584387.2 | XP_005584444.1 | 3 - 4 | 771 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
