Assay Details
| Gene Symbol: RBMX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594570.2 | XP_005594627.1 | 2 - 3 | 229 | 142 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBMX2| Gene Symbol: RBMX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594570.2 | XP_005594627.1 | 2 - 3 | 229 | 142 | |
Set Membership: |
|