Assay Details
| Gene Symbol: MOB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575566.2 | XP_005575623.1 | 5 - 6 | 861 | 147 | |
| XM_005575567.2 | XP_005575624.1 | 5 - 6 | 858 | 147 | ||
| XM_015433532.1 | XP_015289018.1 | 5 - 6 | 749 | 147 | ||
More Information
Set Membership: |
|

MOB1A
Genomic Map