Assay Details
| Gene Symbol: AOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584306.2 | XP_005584363.1 | 1 | 787 | 82 | |
| XM_005584307.2 | XP_005584364.1 | 1 | 787 | 82 | ||
| XM_015438528.1 | XP_015294014.1 | 1 | 787 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005584306.2, XM_005584307.2, XM_015438528.1 transcripts.
Set Membership: |
|

AOC2
Genomic Map