Assay Details
| Gene Symbol: DYDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565288.2 | XP_005565345.2 | 4 | 652 | 143 | |
| XM_015456117.1 | XP_015311603.1 | 4 | 732 | 143 | ||
More Information
Set Membership: |
|

DYDC2
Genomic Map