Assay Details
Gene Symbol: DNTTIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005542682.2 | XP_005542739.1 | 3 | 1736 | 138 | |
XM_005542683.2 | XP_005542740.1 | 3 | 1642 | 138 | ||
XM_005542684.2 | XP_005542741.1 | 2 | 1625 | 138 | ||
XM_005542685.2 | XP_005542742.1 | 2 | 2852 | 138 | ||
GenBank mRNA | AB170577.1 | - | 2 | 1598 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
