Assay Details
Gene Symbol: CD151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005576810.2 | XP_005576867.1 | 6 - 7 | 754 | 100 | |
XM_005576811.2 | XP_005576868.1 | 7 - 8 | 735 | 100 | ||
XM_005576812.2 | XP_005576869.1 | 7 - 8 | 854 | 100 | ||
XM_005576813.1 | XP_005576870.1 | 7 - 8 | 746 | 100 | ||
XM_005576814.2 | XP_005576871.1 | 6 - 7 | 732 | 100 | ||
XM_005576815.1 | XP_005576872.1 | 6 - 7 | 821 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
