Assay Details
| Gene Symbol: HSD17B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319446.1 | NP_001306375.1 | 5 - 6 | 761 | 93 | |
| XM_015438502.1 | XP_015293988.1 | 4 - 5 | 1396 | 93 | ||
| GenBank mRNA | DQ078720.1 | - | 5 - 6 | 761 | 93 | |
More Information
Set Membership: |
|

HSD17B1
Genomic Map